Cerium trinitrate hexahydrate ≥99.5% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

011329-09 011329-22 011329-A1 011329-36
AA11329-09EA 38.87 USD
AA11329-09 AA11329-22 AA11329-A1 AA11329-36
Cerium trinitrate hexahydrate ≥99.5% (REO, rare earth oxide basis), REacton®
Cerium trinitrate hexahydrate

Separation of cerium from other rare earths, catalyst for hydrolysis of phosporic acid esters


Purity: 99.5% CAS No.: 10294-41-4 Molecular Formula: CeN3O9.6H2O Formula Weight: 434.23 (326.13anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic pH: 3 Solubility: Soluble in water, alcohol, acetone Merck Reference: 14,2000 UN No.: 1477 MDL No.: MFCD00149631


Separation of cerium from other rare earths, catalyst for hydrolysis of phosporic acid esters

Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Boiling Pt: 200
Melting Pt: 39
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1 III
Merck Index: 14,2000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR