Cerium(IV) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ceric oxide, Cerium dioxide
011328-30
011328-18
011328-A1
AA11328-30EA
151
USD
AA11328-30
AA11328-18
AA11328-A1
Cerium(IV) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Cerium(IV) oxide
MDL: MFCD00010933
Purity: 99.9%
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: 5 Micron Powder
Density: 7.132
Appearance: Pale yellow
Odor: Odorless
Melting Point: 2600°
Merck Reference: 14,1989
MDL No.: MFCD00010933
Formula:
CeO₂ Boiling Pt: 3468 °C (1013 hPa) Melting Pt: 2600 °C Density: 7.13 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 Merck Index: 12,02035 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...