Lutetium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lutetium (III) nitrate hydrate

011258-04 011258-09
AA11258-04EA 73.23 USD
AA11258-04 AA11258-09
Lutetium trinitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Lutetium trinitrate hydrate

MDL: MFCD00150421


Purity: 99.99% CAS No.: 100641-16-5 Molecular Formula: LuN3O9.xH2O Formula Weight: 360.98(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: White UN No.: 1477 MDL No.: MFCD00150421

Formula: LuN₃O₉·H₂O
MW: 379 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150421
CAS Number: 100641-16-5
EINECS: 233-241-7
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR