Praseodymium(III,IV) oxide ≥99.5% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
011234-A1
011234-14
011234-22
AA11234-A1EA
934
USD
AA11234-A1
AA11234-14
AA11234-22
Praseodymium(III,IV) oxide ≥99.5% (REO, rare earth oxide basis)
Praseodymium(III,IV) oxide
MDL: MFCD00011178
Purity: 99.5%
CAS No.: 12037-29-5
Molecular Formula: Pr6O11
Formula Weight: 1021.43
Physical Form: Powder
Melting Point: 2500°
MDL No.: MFCD00011178
Formula:
O₁₁Pr₆ MW: 1021.44 g/mol Melting Pt: 2012 - 2072 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011178 CAS Number: 12037-29-5 EINECS: 234-857-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...