Arsenic(III) oxide ≥99.996% (metals basis excluding Sb), Puratronic® (max. 0.002% Sb)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Arsenic trioxide
011173-18
011173-04
011173-09
AA11173-18EA
325
USD
AA11173-18
AA11173-04
AA11173-09
Arsenic(III) oxide ≥99.996% (metals basis excluding Sb), Puratronic® (max. 0.002% Sb)
Arsenic(III) oxide
MDL: MFCD00003433
Purity: 99.996%
CAS No.: 1327-53-3
Molecular Formula: As2O3
Formula Weight: 197.84
Physical Form: Powder
Density: 3.74
Melting Point: 312.3°
Boiling Point: 465° subl.
Merck Reference: 14,804
UN No.: 1561
MDL No.: MFCD00003433
Formula:
As₂O₃ MW: 197.84 g/mol Boiling Pt: 465 °C (subl.) Melting Pt: 312.3 °C Density: 3.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1,II Merck Index: 12,00844 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...