Zinc sulfate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

011138-09 011138-18
AA11138-09EA 83.38 USD
AA11138-09 AA11138-18
Zinc sulfate hydrate ≥99.999% (metals basis), Puratronic®
Zinc sulfate hydrate

MDL: MFCD00011302


Purity: 99.999% CAS No.: 16788-42-4 Molecular Formula: O4SZn.xH2O Formula Weight: 161.43(anhy) Physical Form: Crystalline powder Appearance: White Odor: Odorless Melting Point: 100° Merck Reference: 14,10159 UN No.: 3077 MDL No.: MFCD00011302

Formula: ZnSO₄·xH₂O
Melting Pt: 100 °C
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 16788-42-4
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR