Sodium hexachloropalladate(IV) hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

011039-03 011039-06
AA11039-03EA 223 USD
AA11039-03 AA11039-06
Sodium hexachloropalladate(IV) hydrate ≥99.9% (metals basis)
Sodium hexachloropalladate(IV) hydrate

MDL: MFCD00014235


Purity: 99.9% CAS No.: 53823-60-2 Molecular Formula: Cl6Na2Pd.xH2O Formula Weight: 365.10(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red to brown MDL No.: MFCD00014235

Formula: Cl₆Na₂Pd·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00014235
CAS Number: 53823-60-2
EINECS: 258-809-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR