Hydrochloric acid ≥33% ≥99.999999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

010990-AK 010990-AU 010990-AC
AA10990-AKEA 747 USD
AA10990-AK AA10990-AU AA10990-AC
Hydrochloric acid ≥33% ≥99.999999% (metals basis)
Hydrochloric acid

MDL: MFCD00011324 Fieser: 1,308 2,215 4,250 5,322 6,283 7,172 8,246 9,239


Purity: 99.999999% CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid, double distilled in quartz Density: 1.19 Appearance: Colorless to pale yellow Odor: Pungent Boiling Point: 81.5-110° Merck Reference: 14,4780 UN No.: 1789 MDL No.: MFCD00011324

Formula: HCl
MW: 36.46
Boiling Pt: 81.5…110 °C
Melting Pt: –30 °C
Density: 1.19
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
Merck Index: 14,4780

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR