Lithium nitrate, anhydrous ≥99.98% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

010985-03 010985-30 010985-18 010985-09
AA10985-03EA 18.3 USD
AA10985-03 AA10985-30 AA10985-18 AA10985-09
Lithium nitrate, anhydrous ≥99.98% (metals basis)
Lithium nitrate

MDL: MFCD00011094


Purity: 99.98% CAS No.: 7790-69-4 Molecular Formula: LiNO3 Formula Weight: 68.95 Physical Form: Crystalline Density: 2.38 Melting Point: 264° Boiling Point: 600° dec. Merck Reference: 14,5536 UN No.: 2722 MDL No.: MFCD00011094

Formula: LiNO₃
Boiling Pt: 600 °C (dec.)
Melting Pt: 264 °C
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 13,05558

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR