Praseodymium(III,IV) oxide ≥99.996% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

010914-06 010914-14
AA10914-06EA 88.6 USD
AA10914-06 AA10914-14
Praseodymium(III,IV) oxide ≥99.996% (REO, rare earth oxide basis), REacton®
Praseodymium(III,IV) oxide

MDL: MFCD00011178


Purity: 99.996% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178

Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2012 - 2072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR