Ammonium metavanadate ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium monovanadate, Ammonium vanadium trioxide, Ammonium vanadium oxide

010902-06 010902-14
AA10902-06EA 90.7 USD
AA10902-06 AA10902-14
Ammonium metavanadate ≥99.995% (metals basis), Puratronic®
Ammonium metavanadate

MDL: MFCD00011430


Purity: 99.995% CAS No.: 7803-55-6 Molecular Formula: NH4VO3 Formula Weight: 116.98 Physical Form: Powder Density: 2.33 Melting Point: 200° dec. Merck Reference: 14,568 UN No.: 2859 MDL No.: MFCD00011430

Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200 °C (dec.)
Density: 2.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1,II
Merck Index: 13,00572

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR