Aluminium ≥99.999% (metals basis), rods, Puratronic®, Diameter 22 mm (0.87 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

010778-BQ 010778-BU
AA10778-BQEA 212.16 USD
AA10778-BQ AA10778-BU
Aluminium ≥99.999% (metals basis), rods, Puratronic®, Diameter 22 mm (0.87 in)
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈10.35g/cm Odor: Odorless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR