Zinc ≥99.999% (metals basis), shot 1-5 mm (0.04-0.2 in)
Supplier: Thermo Scientific Chemicals
010759-18
010759-30
010759-A1
AA10759-18EA
89.6
USD
AA10759-18
AA10759-30
AA10759-A1
Zinc ≥99.999% (metals basis), shot 1-5 mm (0.04-0.2 in)
Zinc
MDL: MFCD00011291
Purity: 99.999%
CAS No.: 7440-66-6
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...