Trilithium orthophosphate ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
010743-14
010743-22
AA10743-14EA
176
USD
AA10743-14
AA10743-22
Trilithium orthophosphate ≥99.99% (metals basis), Puratronic®
Trilithium orthophosphate
MDL: MFCD00016187
Purity: 99.99%
CAS No.: 10377-52-3
Molecular Formula: Li3PO4
Formula Weight: 115.79
Physical Form: Powder
Density: 2.54
Appearance: White
Odor: Odorless
Melting Point: 837°
MDL No.: MFCD00016187
Formula:
Li₃O₄P Melting Pt: 837 °C Density: 2.54 Storage Temperature: Ambient |
MDL Number:
MFCD00016187 CAS Number: 10377-52-3 EINECS: 233-823-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...