Lithium nitrate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Lithium nitrate, anhydrous is used in the manufacture of red-colored fireworks and flares. has been proposed as a medium to store heat collected from the sun for cooking. A Fresnel lens would be used to melt solid lithium nitrate, which would then function as a solar battery, allowing heat to be redistributed later by convection
Purity: 99.999%
CAS No.: 7790-69-4
Molecular Formula: LiNO3.xH2O (x~0.5)
Formula Weight: 68.95(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,5536
UN No.: 2722
MDL No.: MFCD00011094
Formula:
LiNO₃·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00011094 CAS Number: 7790-69-4 EINECS: 232-218-9 UN: 2722 ADR: 5.1,III Merck Index: 14,5536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...