Lead(II) sulfate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

010730-09 010730-18
AA10730-09EA 78.66 USD
AA10730-09 AA10730-18
Lead(II) sulfate ≥99.999% (metals basis), Puratronic®
Lead(II) sulphate

MDL: MFCD00011166


Purity: 99.999% CAS No.: 7446-14-2 Molecular Formula: PbSO4 Formula Weight: 303.25 Physical Form: Crystalline Density: 6.2 Melting Point: 1170° Merck Reference: 14,5420 UN No.: 2291 MDL No.: MFCD00011166

Formula: O₄PbS
Melting Pt: 1000 °C
Density: 6.0631 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011166
CAS Number: 7446-14-2
EINECS: 231-198-9
UN: 2291
ADR: 6.1,III
Merck Index: 12,05444

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR