Rhenium ≥99.997% (metals basis), powder, Puratronic® -22 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
010673-03
010673-06
AA10673-03EA
103
USD
AA10673-03
AA10673-06
Rhenium ≥99.997% (metals basis), powder, Puratronic® -22 mesh
Rhenium
MDL: MFCD00011195
Purity: 99.997%
CAS No.: 7440-15-5
UN No.: 3089
MDL No.: MFCD00011195
Formula:
Re MW: 186.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...