Zinc ≥99.98% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
010436-BW
010436-BS
010436-CB
AA10436-BWEA
95.62
USD
AA10436-BW
AA10436-BS
AA10436-CB
Zinc ≥99.98% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Zinc
MDL: MFCD00011291
Purity: 99.98%
CAS No.: 7440-66-6
Physical Form: ≈53g/30x10cm
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Specification Test Results
Appearance (Color) | Gray |
Form | Foil |
Comment | 0.22 - 0.28 mm thick |
Width | 27 - 33 mm |
Total Metal Impurities | 0.02% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...