Tin ≥99.8% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

010379-22 010379-36 010379-A3
AA10379-22EA 66.5 USD
AA10379-22 AA10379-36 AA10379-A3
Tin ≥99.8% (metals basis), powder -325 mesh
Tin

MDL: MFCD00133862


Purity: 99.8% CAS No.: 7440-31-5 Physical Form: Apparent density 3.2g/cm3 Odor: Odorless UN No.: 3089 MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
UN: 3089
ADR: 4.1 III
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR