Tantalum ≥99.95% (metals basis), foil, Thickness 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
010356-FI
010356-FF
010356-GH
AA10356-FIEA
350
USD
AA10356-FI
AA10356-FF
AA10356-GH
Tantalum ≥99.95% (metals basis), foil, Thickness 1.0 mm (0.04 in)
Tantalum
MDL: MFCD00011252
Purity: 99.95%
CAS No.: 7440-25-7
Physical Form: ≈10.4g/25x25mm
Odor: Odorless
MDL No.: MFCD00011252
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...