Tantalum ≥99.95% (metals basis), wire, annealed, Diameter 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

010350-G1 010350-G5 010350-G9
AA10350-G1EA 101 USD
AA10350-G1 AA10350-G5 AA10350-G9
Tantalum ≥99.95% (metals basis), wire, annealed, Diameter 1.0 mm (0.04 in)
Tantalum

MDL: MFCD00011252


Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈13.0g/m, Temper: soft Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR