Niobium ≥99.8% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
010275-36
010275-14
010275-22
010275-88
AA10275-36EA
735
USD
AA10275-36
AA10275-14
AA10275-22
AA10275-88
Niobium ≥99.8% (metals basis), powder -325 mesh
Niobium
MDL: MFCD00011126
Purity: 99.8%
CAS No.: 7440-03-1
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011126
Formula:
Nb Storage Temperature: Ambient |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...