Molybdenum ≥99.9% (metals basis), powder -250 mesh

Supplier: Thermo Scientific Chemicals
Warning

000932-30 000932-06
AA00932-30EA 114 USD
AA00932-30 AA00932-06
Molybdenum ≥99.9% (metals basis), powder -250 mesh
Molybdenum

MDL: MFCD00003465


Purity: 99.9% CAS No.: 7439-98-7 Appearance: Grey Odor: Odorless UN No.: 3089 MDL No.: MFCD00003465

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1 III
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR