Lead ≥99.9% (metals basis), bar ∼ 1.35x2.54x30.5 cm (0.53x1.0x12.0 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
000930-KF
000930-KJ
AA00930-KFEA
200
USD
AA00930-KF
AA00930-KJ
Lead ≥99.9% (metals basis), bar ∼ 1.35x2.54x30.5 cm (0.53x1.0x12.0 in)
Lead
MDL: MFCD00134050
Purity: 99.9%
CAS No.: 7439-92-1
Physical Form: ≈1300g/bar
Appearance: Grey
Odor: Odorless
MDL No.: MFCD00134050
Formula:
Pb Boiling Pt: 1740 °C (1013 hPa) Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...