Tantalum ≥99.9% (metals basis), foil, Thickness 0.7 mm (0.03 in)

Supplier: Thermo Scientific Chemicals

000911-FG 000911-FY
AA00911-FGEA 137 USD
AA00911-FG AA00911-FY
Tantalum ≥99.9% (metals basis), foil, Thickness 0.7 mm (0.03 in)
Tantalum

MDL: MFCD00011252


Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈14.5g/25x50mm Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR