Tin ≥99.998% (metals basis), rods, Puratronic®, Diameter 13 mm (0.51 in)

Supplier: Thermo Scientific Chemicals

000788-14 000788-06
AA00788-14EA 328.69 USD
AA00788-14 AA00788-06
Tin ≥99.998% (metals basis), rods, Puratronic®, Diameter 13 mm (0.51 in)
Tin

MDL: MFCD00133862


Purity: 99.998+% CAS No.: 7440-31-5 Physical Form: ≈0.52cm/5g Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR