Yttrium ≥99.5% (REO, rare earth oxide basis), foil, Thickness: 0.64 mm (0.025 in)
Supplier: Thermo Scientific Chemicals
000616-FF
000616-FI
AA00616-FFEA
266
USD
AA00616-FF
AA00616-FI
Yttrium ≥99.5% (REO, rare earth oxide basis), foil, Thickness: 0.64 mm (0.025 in)
Yttrium
Purity: 99.5%
CAS No.: 7440-65-5
Physical Form: ≈1.8g/25x25mm, Packaged in mylar under argon
Odor: Odorless
MDL No.: MFCD00011468
Formula:
Y Boiling Pt: 2927 °C (1013 hPa) Melting Pt: 1525 °C Density: 4.472 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 Merck Index: 12,10240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...