Erbium ≥99.9% (REO, rare earth oxide basis), pieces

Supplier: Thermo Scientific Chemicals
Danger

000111-09 000111-18
AA00111-09EA 135 USD
AA00111-09 AA00111-18
Erbium ≥99.9% (REO, rare earth oxide basis), pieces
Erbium

MDL: MFCD00010987


Purity: 99.9% CAS No.: 7440-52-0 Physical Form: Packaged under argon in glass bottle Appearance: Silvery grey UN No.: 3178 MDL No.: MFCD00010987

Formula: Er
MW: 167.26
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1
UN: 3178
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR