Cerium ≥99.8% (REO, rare earth oxide basis), ingot
Supplier: Thermo Scientific Chemicals
|
Danger
|
000065-18
000065-30
AA00065-18EA
159
USD
AA00065-18
AA00065-30
Cerium ≥99.8% (REO, rare earth oxide basis), ingot
Cerium
MDL: MFCD00010924
Purity: 99.8% min
CAS No.: 7440-45-1
Physical Form: Packaged under mineral oil
Appearance: Grey
Odor: Odorless
UN No.: 1333
MDL No.: MFCD00010924
Formula:
Ce Boiling Pt: 3468 °C (1013 hPa) Melting Pt: 795 °C Density: ~ 6.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010924 CAS Number: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...