Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99.0%, Biotechnology Grade 1 Phase, VWR®

Supplier: VWR
Danger

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

K169-400ML K169-100ML
97064-826CS 1574.98 USD
97064-826 97064-824
Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99.0%, Biotechnology Grade 1 Phase, VWR®
Phenol : Chloroform : iso-Amyl alcohol
Storage Temperature: Ambient
MDL Number: MFCD00133763
CAS Number: 136112-00-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Conductivity(50%, Water)@25°C 250 uS - 330 uS
pH(50%, Water)@25°C 8.05 - 8.35

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR