Acetonitrile, anhydrous 99,8, ChemSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Argon_Chemseal

043166-K2 043166-AK
AA43166-K2EA 205.27 USD
AA43166-K2 AA43166-AK
Acetonitrile, anhydrous 99,8, ChemSeal™
Acetonitrile

Packaged under argon in resealable ChemSeal bottle


Purity: 99.8+% CAS No.: 75-05-8 Molecular Formula: CH3CN Formula Weight: 41.05 Flash Point: 5°(41°F) Physical Form: Liquid Density: 0.786 Refractive Index: 1.3440 Odor: Pungent Melting Point: -48 to -45° Boiling Point: 80-82° Beilstein Reference: 741857 Merck Reference: 14,70 UN No.: 1648 MDL No.: MFCD00001878

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.786
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 14,70
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR