Uridine

Supplier: VWR

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dioneUracil-1-β-D-ribofuranoside1-β-D-Ribofuranosyluracil

0975-50G
97062-542CS 2132.57 USD
97062-542
Uridine
Uridine
Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 166 °C…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00006526
CAS Number: 58-96-8
EINECS: 200-407-5

Order Now

Specification Test Results

Em (261nm, 0.15M Phosphate Buffer, pH 7.3) = 10100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR