Potassium hydroxide, Reagent Grade

Supplier: VWR
Danger

Synonyms: Potash caustic, Caustic potash

0489-500G 0489-2.5KG 0489-1KG
97064-598CS 556.24 USD
97064-598 97064-596 97064-988
Potassium hydroxide, Reagent Grade
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Arsenic <= 0.0003%
Heavy Metals <= 0.003 %
Moisture 10 - 17 %
Potassium Carbonate <= 3.5 %
Purity >85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR