Clarity One-Step Immuno Fecal Occult Blood Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-FOB01
DTG-IFOB04
95064-042CS
6293.92
USD
95064-042
10154-006
Clarity One-Step Immuno Fecal Occult Blood Test, Clarity Diagnostics
Clinical Diagnostic Supplies
Clinical Reagents and Kits
Early detection of colorectal cancer in patients can be critical for successful treatment. Clarity has developed a product that makes the testing process easier and gives you clear results. Introducing Clarity’s One-step Immuno Occult Blood Test Kit or IFOBT for short.
- No food, vitamin, or iron restrictions
- Greater sensitivity (0.05ug hHB/ml monoclonal antibody)
- Room temperature storage
- Contains external controls (DTG-FOBCTLS)
- 28 month dating
- 100% Medicare reimbursement
With built-in quality control.
Certificaciones: CLIA waived.