hCG Pregnancy Strips and Cassettes, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-PLUS25 DTG-PLUS50 DTG-HCG25 DTG-HCG50 DTG-HCG100 DTG-COMBO
95064-021CS 1371.19 USD
95064-021 95064-023 95064-133 95064-135 95064-137 95064-145
hCG Pregnancy Strips and Cassettes, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Open a new revenue stream by bringing your pregnancy testing in-house with Clarity’s array of Pregnancy Testing Products. Clarity offers hCG Urine Cassettes, hCG Combo (urine/serum) Cassettes, hCG Urine Strips, and hCG/Urine Controls that are high-quality and at a value that is unmatched by competitors.
  • Available in strips and cassettes
  • Detects pregnancy first day of missed period
  • Cassettes only require 3 drops of urine sample
  • Sensitivity of 25 mIU/mL, specificity of 100%
  • FDA cleared and CLIA waived
  • No cross reactivity with FSH, LH, and TSH
Clarity’s tests are fast and reliable, designed to save your practice time and money while helping to build your reputation as an efficient reliable practice in the minds of your patients.
Certificaciones: FDA approved.