Vaprox® Hydrogen Peroxide Sterilants, 35%
Supplier: Steris
PB006US PB027US
94024-562CS 1178.13 USD
94024-562 94024-564
Vaprox® Hydrogen Peroxide Sterilants, 35%
Cleaning Agents Disinfectants
Proprietary 35% aqueous hydrogen peroxide solution that’s compatible with a wide range of environmental surfaces and offers broad-spectrum efficacy against viruses, bacteria, yeasts, and bacterial spores while leaving no toxic residue.
  • Performance - Formulated and tested for optimum VHP Biodecontamination Unit operation and reliability; Wide range of material compatibility
  • Efficacy - Broad spectrum anti-microbial; Validated biodecontamination cycles for use on environmental surfaces
  • Quality- Proper testing of incoming raw material and each manufactured lot; Manufactured to lot control in accordance with cGMP procedures; Filtered during manufacturing for optimal Biodecontamination Unit operation
  • Productivity - Containers offered in a variety of sizes from cups through 5 gallons to accommodate application requirements
  • Standards - US EPA registration No. (EPA Reg. No. 58779-4)