KLEENGUARD® A70 Chemical Spray Protection Smocks, Kimberly-Clark Professional®
Supplier: Ansell Healthcare
9830 9829
89065-964CS 439 USD
89065-964 89065-962
KLEENGUARD® A70 Chemical Spray Protection Smocks, Kimberly-Clark Professional®
Disposable Garments
These chemical spray protection smocks are constructed of a 1.5mil polyethylene film-coated, spunbond polypropylene fabric.
  • Seamless design
  • Soft and durable fabric, and a wick-away lining for increased comfort
  • Elastic wrists, tie front, and open back
  • NFPA 99 compliant for antistatic material
The KLEENGUARD® A70 line of apparel is designed to provide protection against chemical spray in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications.