Accessories for Rotors, Beckman Coulter

Supplier: Beckman Coulter
392805 392806 392873 393070
BK392805PK 286.08 USD
BK392805 BK392806 BK392873 BK393070
Accessories for Rotors, Beckman Coulter
Centrifuges Rotors
This assembly is compatible with Allegra® X-12, X-14, and X-15R series centrifuges (BK392472, BKA99464, and BK392932 series).

  • Application: Rapid Sediment of Protein Precipitates, Large Particles, Cells and Cell Debris
  • Material: Aluminum and Stainless Steel
  • Maximum Speed: 4450 rpm
  • Rubber Pads and Carriers Included

Each rotor has carriers that accommodate up to three standard microplates: one Beckman Coulter deep-well plate, one Beckman Coulter square-well plate, or one 96-tube rack.

It provides g forces up to 2890 g (3750 rpm) in Allegra X-12 and up to 4060 g (4450 rpm) in Allegra X-15R centrifuges for cellular assays and quick microplate spins.

Certificaciones: BioCertified covers are certified by CAMR, Porton Down, UK for biocontainment.

Información para pedidos: Each microplate carrier assembly includes two carriages and two rubber pads.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR