CLP–Semi-Volatiles Acid Surrogate Standard (3/90 SOW), Restek
Supplier: RESTEK CORP
31083 31073 31003
RK31083EA 175.5 USD
RK31083 RK31073 RK31003
CLP–Semi-Volatiles Acid Surrogate Standard (3/90 SOW), Restek
Standards Environmental Analysis Standards
Standards are packaged in methanol at the stated concentration in an ampoule.
  • Mixture contains 2-Chlorophenol-d4, 2-Fluorophenol, Phenol-d6, and 2,4,6-Tribromophenol
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).