Base/Neutrals Matrix Spike Standards, Restek
Supplier: RESTEK CORP
31004 31074 31084 31004-15
RK31004EA 43.5 USD
RK31004 RK31074 RK31084 75834-594
Base/Neutrals Matrix Spike Standards, Restek
Standards Environmental Analysis Standards
Standards are packaged in methanol in an ampule.
  • Highest concentrations commercially available
  • Reduces laboratory cost per sample extract
  • Mixture contains Acenaphthene, 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, N-Nitroso-di-n-propylamine, Pyrene, and 1,2,4-Trichlorobenzene
Each standard includes an ampule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).