VWR® Phosphate Buffered Saline (PBS)
Supplier: VWR International
SH3A12506.01 SH3A12506.LS
77661-370EA 28.8 USD
77661-370 77661-372
VWR® Phosphate Buffered Saline (PBS)
Buffers Cell Culture Buffers
Balanced salt solution used for multiple cell culture applications.
  • Full traceability and documented origin of all formula ingredients
  • Validated and consistent production processes
0.1 µm sterile filtered
Endotoxin concentration: ≤1.0 EU/ml
Certificaciones: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.