InhibiTaq™ 2X HotStart qPCR MasterMix
Supplier: EMPIRICAL BIOSCIENCE
IT-MM-200 IT-MM-600
77676-302EA 123.38 USD
77676-302 77676-304
InhibiTaq™ 2X HotStart qPCR MasterMix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
2X InhibiTaq™ hot start qPCR master mix is a 2X ready-to-use master mix for quantitative real time evaluation of DNA using dye- or fluorescent probe-based detection. InhibiTaq™ hot start qPCR master mix contains a chemically modified taq DNA polymerase that remains completely inactive at room temperature. The enzyme becomes activated after only 2 minutes at 95 °C.
  • Fast qPCR
  • Fast reverse transcription PCR/qPCR
  • Inhibitor tolerant qPCR
  • Inhibitor tolerant reverse transcription PCR/qPCR, qPCR (probe)
  • cGMP manufacturing
InhibiTaq™ hot start qPCR master mix has been optimized for use with hydrolysis-based probes but is also suitable with other probe-based detection systems and dye-based detection.
Certificaciones: ISO 13485.