InhibiTaq™ 4X PLUS Hot Start qPCR MasterMix
Supplier: EMPIRICAL BIOSCIENCE
ITP-MM-200 ITP-MM-600
77676-310EA 139.79 USD
77676-310 77676-312
InhibiTaq™ 4X PLUS Hot Start qPCR MasterMix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
An enhanced performance MasterMix containing UDG Enzyme and dUTP to minimize the effects of carry over contamination. The MasterMix is a 4X ready-to-use MasterMix for quantitative real time evaluation of DNA using dye- or fluorescent probe-based detection.
  • Applications: Carryover prevention, Fast qPCR, Fast reverse transcription PCR/qPCR, Inhibitor tolerant qPCR, Inhibitor tolerant reverse transcription PCR/qPCR, qPCR (probe)
  • Availability: cGMP Manufacturing
InhibiTaq™ PLUS HotStart qPCR MasterMix contains a chemically modified Taq DNA polymerase that remains completely inactive at room temperature. The enzyme becomes activated after only 2 minutes at 95 °C. Empirical's InhibiTaq™ PLUS HotStart qPCR MasterMix has been optimized for use with hydrolysis-based probes but is also suitable with other probe-based detection systems and dye-based detection.
Certificaciones: ISO 13485 certified.