Aquasoft Nitrile Exam Gloves
Supplier: O&M HALYARD, INC
43932 43933 43934 43935 43936
77749-884BX 29.58 USD
77749-884 77749-886 77749-888 77749-890 77749-892
Aquasoft Nitrile Exam Gloves
Gloves
Nitrile gloves provides a new level of comfort, protection and value. Tactile sensitivity comparable to latex without the risk of Latex allergies.
  • Exceptional tactile sensitivity
  • Non-sterile
  • 300 count per box to save space and waste
  • Textured fingertips
  • Risk category: Tested - 34 Chemo drugs
These gloves are ideal for applications that require superior tactile sensitivity. They are tested and approved for use against 34 Chemotherapy drugs.
Certificaciones: ASTM D6978, ASTM F739, ASTM D6319, ASTM D5151, ISO 10993-10, ISO 9001.
Envase: 300 Gloves per box, 10 boxes per case, 3000 gloves per case.