T4 DNA Polymerase

Supplier: EMPIRICAL BIOSCIENCE

T4DP-300 T4DP-900
77676-364EA 50.39 USD
77676-364 77676-366
T4 DNA Polymerase
Enzymes

Order Now

SPECIFICATIONS

Environmentally Preferable
Source
Species
Storage Conditions −10...−25 °C
Shipping Conditions 70

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR