MeanGreen™ Loading Dyes, 6X
Supplier: EMPIRICAL BIOSCIENCE
Contains blue and yellow loading dyes and a density agent that allow reactions to be loaded directly onto agarose gels and make monitoring progress during electrophoresis easy.
- For Endpoint PCR application
- Store at −20 °C upon arrival
- Minimize number of freeze thaw cycles by storing in working aliquots
These dyes do not obscure visualization of DNA bands on the gel as the dye fronts run outside the size of most PCR products. The 6X MeanGreen™ Loading Dye is supplied in a solution with Glycerol, Tris-HCl, and EDTA.
Certificaciones: ISO 13485.