2X qPCR Probe MasterMix with Separate ROX

Supplier: EMPIRICAL BIOSCIENCE

QP-MM-1000 QP-MM-500
77676-336EA 328.72 USD
77676-336 77676-338
2X qPCR Probe MasterMix with Separate ROX
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits

qPCR Probe MasterMix: Supplied as a 2X ready-to-use MasterMix for quantitative real time evaluation of DNA using fluorescent probe-based detection.


  • Applications: qPCR (probe)
  • Storage and handling: Store at −20 °C upon arrival


It contains flashtaq HotStart DNA polymerase, a chemically modified taq DNA polymerase that remains completely inactive at room temperature. The enzyme becomes activated after only 2 minutes at 95 °C.


It includes dNTPs, MgCl₂, and optimized buffer for fast, efficient qPCR. Empirical's qPCR probe MasterMix has been optimized for use with hydrolysis-based probes such as TaqMan but is also suitable with other probe-based detection systems.


Certificaciones: ISO 13485.


Información de suministro: 25 µm ROX reference dye supplied separately for use with some real time machines.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR