BioReady Gold Nanoshells, 150 nm for Passive Conjugation

Supplier: NANOCOMPOSIX, INC.
GSCR150-30M GSCR150-5M GSCR150-100M
77680-322EA 684 USD
77680-322 77680-324 77680-320
BioReady Gold Nanoshells, 150 nm for Passive Conjugation
Beads Microcarriers
150 nm gold nanoshells for passive adsorption combine strong plasmonic optical properties with surface chemistry designed for passive conjugation. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.

  • Hydrodynamic Ø (DLS): 145 to 195 nm
  • Zeta potential: ≤–35 mV
  • pH of solution: 5.8 to 8.5
  • Peak wavelength (λmax): 730 to 820 nm
  • Solvent: 0.2 mM sodium citrate

BioReady Gold Nanoshells enable enhanced sensitivity in lateral flow assays (LFA) due to their intense optical signal. Integration of these nanoparticles into LFAs can provide increased signal-to-noise ratios and lowered limits of detection for your target of interest.

Certificaciones: ISO 13485/cGMP.

Precaución: Intended for research use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR