Abgene™ Caps and Cap Strips
Supplier: Thermo Fisher Scientific
AB-1179 AB-0981
77753-642CS 261.3 USD
77753-642 77753-670
Abgene™ Caps and Cap Strips
Seals Microplate Seals
Thermo Scientific™ Abgene™ caps and cap strips are suitable for round well plates and feature a low-profile for increased well volume.
  • Eight cap strip or individual caps
  • Low profile for increased well volume
  • End tabs for easy removal
  • Cap strip caps are individually numbered
  • hDNA-free, DNase-/RNase-free, and PCR inhibitor-free
Compatible plates: 77753-652, 77753-660, 77753-698, 77753-702, 77753-708.