Anti-CALD1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43565-50UL A43565-100UL
77706-121EA 228.38 USD
77706-121 77706-120
Anti-CALD1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:150

Type:
Antigen: CALD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR